Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pilot® G-Knock Be Green® Retractable Roller Ball Pen

Supplier: Janitorial Supplies
Pilot® Be Green®
PIL31506 PIL31507 PIL31508
Gel ink pen is made from 81.5% recycled material, making saving the planet just a little easier. These roller ball pens are refillable. Made with Pocket clip.

Dynamic, smear-proof gel ink formula offers clean, smooth writing. Retractable style protects your pockets and purses. Comfortable, contoured rubber grip ensure a pleasant writing experience.



 

ORDER

Expand All / Collapse All
Ink Color Point Size Tip Type Barrel Color Refill Number Point Type Supplier No. VWR Catalog Number Unit Price Quantity
Blue 0.7 mm Conical Translucent PIL77241 Fine PIL31507 500020-647 Dozen Retrieving
Product and/or packaging contains recycled/renewable content
Black 0.7 mm Conical Translucent PIL77240 Fine PIL31506 500035-476 Dozen Retrieving
Product and/or packaging contains recycled/renewable content
Red 0.7 mm Conical Translucent PIL77242 Fine PIL31508 500035-478 Dozen Retrieving
Product and/or packaging contains recycled/renewable content
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty