Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wausau Paper® Vellum Bristol Cover Stock

Supplier: Janitorial Supplies
Wausau Paper®
WAU49581 WAU82341 WAU82351 WAU82361 WAU82321 WAU82331
Durable semi-smooth cover for mediumweight to heavyweight applications: files, color-coding, posters and covers.

Laser and inkjet printer guaranteed.



 

ORDER

Expand All / Collapse All
Dimensions Color Weight Supplier No. VWR Catalog Number Unit Price Quantity
81/2 x 11" Blue 67 lbs. WAU82321 500018-806 Pack of 250 Retrieving
81/2 x 11" Green 67 lbs. WAU82351 500018-807 Pack of 250 Retrieving
81/2 x 11" Gray 67 lbs. WAU82341 500018-808 Pack of 250 Retrieving
81/2 x 11" Ivory 67 lbs. WAU82361 500018-809 Pack of 250 Retrieving
81/2 x 11" Canary Yellow 67 lbs. WAU82331 500018-811 Pack of 250 Retrieving
81/2 x 11" Ivory 110 lbs. WAU49581 500040-432 Pack of 250 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty