Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HON® Slide-Away Keyboard Platform

Supplier: Janitorial Supplies
HON®
HON4022N HON4022C
Strong, durable laminate helps resist against scratches and stains

Can be positioned 2" to 3-1/2" below worksurface. Holds keyboards up to 21-1/2w x 10d.



 

ORDER

Expand All / Collapse All
Color Keyboard Tray Dimensions (W x D) Height (+/- Desktop) Workstation Type Material Glide Track Length 360° Swivel Mouse Tray Swivels Drawer Slide Mechanism Removable Mouse Tray Height Adjustment Sit/Stand Frame Sliding Tray Tilt Adjustable Wrist Rest Included For Use With Supplier No. VWR Catalog Number Unit Price Quantity
Harvest 21 1/2" x 10" 2" to 3 1/2" Straight Laminated - No No Slide-Away No No No Yes No No HON® 10500 Series Workstation Return or Curved Corner Workstation, HON® 10600 Series Kneespace Credenza or Workstation Return, HON® 10700 Series Kneespace or Single Pedestal Credenza, HON® 94000 Series Kneespace Credenza or Return HON4022C 500043-040 Each Retrieving
Product and/or packaging is safer to human and environmental health
Mahogany 21 1/2" x 10" 2" to 3 1/2" Straight Laminated 17" No No Slide-Away No No No Yes No No HON® 10500 Series Workstation Return or Curved Corner Workstation, HON® 10600 Series Kneespace Credenza or Workstation Return, HON® 10700 Series Kneespace or Single Pedestal Credenza, HON® 94000 Series Kneespace Credenza or Return HON4022N 500021-487 Each Retrieving
Product and/or packaging is safer to human and environmental health
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging is safer to human and environmental health
Environmental Certification Scientific Certification Systems (SCS) Certified Indoor Advantage™

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty