Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pendaflex® Six-Section PressGuard® Colored Classification Folders

Supplier: Janitorial Supplies
Pendaflex® PressGuard®
PFX1257GR PFX2257Y PFX2257SC PFX1257Y PFX1257SC PFX2257GR PFX2257BL PFX1257LB PFX2257LB PFX1257BL
A bright idea for color-coding and subdividing files

Features acrylic-coated 20 pt. PressGuard® covers that resist stains and moisture. 2" capacity fasteners on the interior covers. Two-way 1" capacity fasteners on the two 17 pt. Kraft dividers for further classifying. Bonded fasteners resist catching on loose paper. Color-matched, reinforced rip proof tape gusset delivers durable expansion. 2/5 cut top tabs in right of center position.



 

ORDER

Expand All / Collapse All
Size Color Expansion Supplier No. VWR Catalog Number Unit Price Quantity
Legal Blue 2" PFX2257BL 500011-477 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Legal Light Blue 2" PFX2257LB 500011-480 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Legal Green 2" PFX2257GR 500011-479 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Letter Light Blue 2" PFX1257LB 500011-499 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Letter Scarlet 2" PFX1257SC 500011-496 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Letter Green 2" PFX1257GR 500011-498 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Letter Blue 2" PFX1257BL 500011-497 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Letter Yellow 2" PFX1257Y 500011-444 Box of 10 Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste
No. of sections 6
No. of Fasteners 6
Environmental Certification Sustainable Forestry Initiative® (SFI) Certified

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty