Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bush® Series A Hutch

Supplier: Janitorial Supplies
Bush®
BSHWC94473 BSHWC57473 BSHWC57461 BSHWC94461 BSHWC57437 BSHWC94437 BSHWC94449 BSHWC57449
Contemporary laminate hutch makes a professional statement in any office

Vacu-form, vinyl-clad doors for added durability. Fabric-covered tackboard holds memos, notes and photos. Finished back panel adds privacy and allows hutch to be used as a divider between workstations. Self-closing adjustable hinges for precise operation.



 

ORDER

Expand All / Collapse All
Material For Use With Depth Height Width Color No. of Compartments No. of Doors Compartment Depth Compartment Height Compartment Width Hutch Special Features No. of Compartments (text) Supplier No. VWR Catalog Number Unit Price Quantity
Laminated Light Pack, Series A 72" Wide Workstation Desk 13 7/8 " 36 1/2 " 72 " Galaxy, Hansen Cherry - 4 11.77" 12.99" 12.84", 27.54" 72" Wide 1 Open, 2 Enclosed BSHWC94473 500013-615 Each Retrieving
Laminated Light Pack, Series A 36" Wide Workstation Desk 13 7/8 " 36 1/2 " 36 " Natural Cherry, Slate Gray - 2 11 7/8" 13" 13 3/8" 36" Wide 1 Open, 2 Enclosed BSHWC57437 500013-606 Each Retrieving
Laminated Light Pack, Series A 36" Wide Workstation Desk 13 7/8 " 36 1/2 " 36 " Galaxy, Hansen Cherry - 2 11 7/8" 13" 13 3/8" 36" Wide 1 Open, 2 Enclosed BSHWC94437 500013-607 Each Retrieving
Laminated Light Pack, Series A 48" Wide Workstation Desk 13 7/8 " 36 1/2 " 48 " Galaxy, Hansen Cherry - 2 11.89", 11.97" 12.99" 13.45", 17.01" 48" Wide 1 Open, 2 Enclosed BSHWC94449 500013-609 Each Retrieving
Laminated Light Pack, Series A 60" Wide Workstation Desk 13 7/8 ", 13 1/2 " 36 1/2 " 60 " Natural Cherry, Slate Gray 2 4 11 7/8" 13" 27 7/8" 60" Wide - BSHWC57461 500013-610 Each Retrieving
Laminated Light Pack, Series A 72" Wide Workstation Desk 13 7/8 " 36 1/2 " 72 " Natural Cherry, Slate Gray - 4 11.77" 12.99" 12.84", 27.54" 72" Wide 1 Open, 2 Enclosed BSHWC57473 500013-616 Each Retrieving
Laminated Light Pack, Series A 48" Wide Workstation Desk 13 7/8 " 36 1/2 " 48 " Natural Cherry, Slate Gray - 2 11.89", 11.97" 12.99" 13.45", 17.01" 48" Wide 1 Open, 2 Enclosed BSHWC57449 500013-608 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty