Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avery® Removable Self-Adhesive ID Labels

Supplier: Janitorial Supplies
Avery®
AVE6460 AVE6467 AVE6465 AVE6464
Create and print your own labels.

  • Rectangular shape

Labels stick, stay and remove easily.



 

ORDER

Expand All / Collapse All
Material For Use With Dimensions Color Packaging Shape Supplier No. VWR Catalog Number Unit Price Quantity
Paper Inkjet Printers, Laser Printers 1/2 x 13/4" White 80/Sheet, 2000/Unit, 25 Sheets/Unit Rectangular AVE6467 500016-039 Pack of 2,000 Retrieving
Paper Inkjet Printers, Laser Printers 1 x 25/8" White 30/Sheet, 750/Unit, 25 Sheets/Unit Rectangular AVE6460 500016-036 Pack of 750 Retrieving
Paper Inkjet Printers, Laser Printers 31/3 x 4" White 6/Sheet, 150/Unit, 25 Sheets/Unit Rectangular AVE6464 500016-037 Pack of 150 Retrieving
Paper Inkjet Printers, Laser Printers 81/2 x 11" White 1/Sheet, 25/Unit, 25 Sheets/Unit Rectangular AVE6465 500016-038 Pack of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty