Would you like to log in again?
Tennsco Regal Shelving Starter Set and Add-On Unit
Supplier: Janitorial Supplies
Tennsco
TNNRGL1836ASD TNNRGL1836SSD TNNRGL1236ASD TNNRGL1536SSD TNNRGL1536ASD TNNRGL1236SSD
Heavy-duty rolled steel, sand color frame with oak laminate shelves that adjust every 1-1/2"
Over 600-lb. capacity per shelf. Each Starter Set and Add-On Unit includes five adjustable shelves, top shelf and 12 shelf supports.
ORDER
Expand All / Collapse All
Material Depth Height Width Color No. of Shelves Adjustability Overall Unit Weight Capacity Shelf Capacity (Weight) Supplier No. VWR Catalog Number Unit Price Quantity
Heavy-Duty Rolled Steel 12 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1236ASD 500024-778 Each Retrieving
Heavy-Duty Rolled Steel 12 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1236SSD 500024-774 Each Retrieving
Heavy-Duty Rolled Steel 15 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1536SSD 500024-775 Each Retrieving
Heavy-Duty Rolled Steel 18 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1836SSD 500024-776 Each Retrieving
Heavy-Duty Rolled Steel 15 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1536ASD 500024-779 Each Retrieving
Heavy-Duty Rolled Steel 18 " 76 " 36 " Sand 6 5 Adjustable Shelves, Shelves Adjust In 1 1/2" Increments 3600 lbs. 600 lbs. TNNRGL1836ASD 500024-780 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty