Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Apollo® Projection & Microfilm Replacement Lamp

Supplier: Janitorial Supplies
Apollo®
APOAEVD APOAEVW APOAFXL APOAEYB APOADYS APOAENX
Long life and accurate color 99% pure quartz glass for brightest light.



 

ORDER

Expand All / Collapse All
Voltage Application For Use With Supplier No. VWR Catalog Number Unit Price Quantity
120.0 V Projectors Buhl Travelling Overhead Projectors, Bell Howell/Eiki Travelling Overhead Projectors, Da-Lite Travelling Overhead Projectors, 3M Travelling Overhead Projectors, Apollo® AI1000 APOADYS 500016-425 Each Retrieving
82.0 V Projectors 3M 4405, Apollo® Al-1000B, 3000, 3002, Apollo® Horizon Educational, Buhl, Da-Lite, Dukane EP1800, For Bell & Howell/Eiki, Ventura 4000 APOAEYB 500016-428 Each Retrieving
82.0 V Projectors 3M Full-Size Overhead Projectors, 9000 Series,1810, 1880, 1720, Apollo® Concept Series, Buhl Overhead Projectors, Da-Lite/Besseler, Dukane 653, Elmo 4327, For Apollo® Eclipse, Al-2000 & Al-2004 APOAENX 500016-427 Each Retrieving
82.0 V Projectors Apollo Eclipse/Concept; Odyssey; Dukane; 3M Products APOAFXL 500016-429 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty