Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TOPS® Prism™ + Colored Writing Pads

Supplier: Janitorial Supplies
TOPS® Prism™
TOP63016 TOP63116 TOP63190 TOP63090 TOP63030 TOP63140 TOP63150 TOP63040 TOP63050 TOP63160 TOP63060 TOP63120 TOP63020 TOP63130
Ruled writing pads with extra-heavy chipboard back.

  • Top binding edge
  • Letr-Trim® perforation
  • Unpunched



 

ORDER

Expand All / Collapse All
Height Width Color Lines Per Page Backing Binding No. of Sheets per Pad Ruling Weight Supplier No. VWR Catalog Number Unit Price Quantity
8" 5" Pastel Blue 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63020 500018-237 Pack of 12 Retrieving
8" 5" Pastel Green 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63090 500018-238 Pack of 12 Retrieving
8" 5" Pastel Gray 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63060 500018-239 Pack of 12 Retrieving
8" 5" Pastel Ivory 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63030 500018-240 Pack of 12 Retrieving
8" 5" Pastel Orchid 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63040 500018-241 Pack of 12 Retrieving
8" 5" Pastel Pink 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63050 500018-242 Pack of 12 Retrieving
8" 5" Assorted Pastels 22 Chipboard, 60 pt Strip 50 Jr. Legal 16 lbs. TOP63016 500018-243 Pack of 6 Retrieving
113/4" 81/2" Pastel Blue 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63120 500018-244 Pack of 12 Retrieving
113/4" 81/2" Pastel Green 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63190 500018-245 Pack of 12 Retrieving
113/4" 81/2" Pastel Gray 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63160 500018-246 Pack of 12 Retrieving
113/4" 81/2" Pastel Ivory 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63130 500018-247 Pack of 12 Retrieving
113/4" 81/2" Pastel Orchid 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63140 500018-248 Pack of 12 Retrieving
113/4" 81/2" Pastel Pink 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63150 500018-249 Pack of 12 Retrieving
113/4" 81/2" Assorted Pastels 28 Chipboard, 60 pt Strip 50 Legal 16 lbs. TOP63116 500018-250 Pack of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty