Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safco® Mobile Laminate Machine Stand With Pullout Drawer

Supplier: Janitorial Supplies
Safco®
SAF1852MH SAF1852CY
If your office is on the move make sure your AV and multimedia can move with you!

  • Scratch resistant, stain resistant top
  • Wheels for mobility and lock for security

Use as a machine stand or beverage cart with fully enclosed storage compartments. Double door cabinet provides concealed storage. Top has 2" high lip on three sides to secure machines or corral spills. Interior shelf adjusts for storing larger items.



 

ORDER

Expand All / Collapse All
Material Capacity Depth Height Width Caster Size Casters Color Door Type Finish No. of Doors No. of Drawers No. of Shelves No. of Wheels Door Type/Features Supplier No. VWR Catalog Number Unit Price Quantity
Furniture Grade Particleboard 200 lbs. 20 1/2" 36 1/4" 30" 2" 4 Swivel Casters (2 Locking) Cherry - Laminate 2 1 1 Adjustable Interior Shelf 4 Black Pulls SAF1852CY 500044-147 Each Retrieving
Product and/or packaging contains recycled/renewable content
Furniture Grade Particleboard 200 lbs. 20 1/2" 36 1/4" 30" 2" 4 Swivel Casters (2 Locking) Mahogany Black Pulls Laminate 2 1 1 Adjustable Interior Shelf 4 - SAF1852MH 500025-377 Each Retrieving
Product and/or packaging contains recycled/renewable content
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content
Storage Capacity Lower Storage Compartment: 28 1/2w x 18 3/4d", Pullout Drawer: 27 1/2w x 17 3/4d x 5h"
Top thickness 3/4"

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty