Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

tools-scrapers

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"5"
Sort by:
 
 
 
 

Catalog Number: (500024-510)
Supplier:  Janitorial Supplies
Description:   This versatile tool is a necessity for painting and comes in handy for a variety of other jobs. Features retractable blade for trimming, cutting and s...
Catalog Number: (500024-509)
Supplier:  Janitorial Supplies
Description:   4" zinc alloy angled head. Removes gum and wax buildup along baseboards or floors with ease. Use the blade’s razor edge for hard floors, the blunt edg...
Supplier:  Janitorial Supplies
Description:   Includes dispenser.
Catalog Number: (500024-511)
Supplier:  Janitorial Supplies
Description:   For scraping windows, walls and floors. Use the razor edge to scrape paint and tape, the blunt edge for cement and stucco. Includes one reversible car...
Supplier:  Janitorial Supplies
Description:   Replacement blade for Unger® heavy-duty scraper. Dual sided. Razor-sharp edge eliminates the need for strenuous scraping on all hard surface floors; b...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 5  of 5