Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "rubber-bands-rubber-bands"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   100% latex-free bands contain no natural rubber proteins and have the added benefit of an antimicrobial agent, which inhibits the growth of fungi, mol...
Supplier:  Janitorial Supplies
Description:   These translucent amber rubber bands are made with a high rubber content to provide added stretch capability and longevity.
Supplier:  Janitorial Supplies
Description:   General purpose rubber bands for home or office use.
Supplier:  Janitorial Supplies
Description:   Excellent, easy stretch to help avoid Carpal Tunnel Syndrome. Perfect for repeated use and fast application.
Catalog Number: (500024-269)
Supplier:  Janitorial Supplies
Description:   Secure files, printouts, catalogs, small can liners and much more. These rubber bands can handle jobs ordinary bands can't. 12 per pack.
Catalog Number: (10746-526)
Supplier:  Janitorial Supplies
Description:   Features the six most popular rubber band sizes in a 1.5 oz. box. All rubber bands are color-coded for quick and easy identification. Assorted colors....
Supplier:  Janitorial Supplies
Description:   Heavy-duty, reusable bands are excellent for stabilizing pallet loads and to bundle, hold or organize as moving bands. Easy-to-use bands are color-cod...
Supplier:  Janitorial Supplies
Description:   100% latex-free bands are made from synthetic rubber and contain no natural rubber proteins to protect users from latex allergy reactions. These bands...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 12
  1  2  Next