Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "office-environment-air-cleaners-filters"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Filtrete air cleaning filters snap easily into place after removing used filters. Replace filter every three months to ensure maximum performance of y...
Catalog Number: (102830-864)
Supplier:  Janitorial Supplies
Description:   High efficiency filter helps remove airborne particles and allergens from indoor air. Carbon filtration reduces unpleasant odors up to 44%. Dual-posit...
Catalog Number: (500022-802)
Supplier:  Janitorial Supplies
Description:   The next generation ionic air purifier. This incredibly effective, totally silent system improves the air quality of the environment by trapping airbo...
Catalog Number: (500043-145)
Supplier:  Janitorial Supplies
Description:   Helps reduce common household odors. In addition, helps maximize the HEPA filter life. Should be replaced approximately every 3 months.
Catalog Number: (500043-146)
Supplier:  Janitorial Supplies
Description:   95% efficient HEPA-type filter. Reduces particles and unpleasant odors. Should be replaced approximately every 6 months.
Supplier:  Janitorial Supplies
Description:   Replacement Carbon Filter for Harmony Air Purifiers, 4/Box
Supplier:  Janitorial Supplies
Description:   Permanent IFD filter captures mold, smoke, dander, dust and pollen. Quietly and efficiently dispenses clean air. Three air cleaning power levels and c...
Catalog Number: (500043-147)
Supplier:  Janitorial Supplies
Description:   Captures 99.97% of microscopic allergens such as mold spores, pollen, dust, smoke and pet dander. Should be replaced once a year.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8