Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "meeting-room-accessories-pa-systems"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
Sort by:
 
 
 
 

Catalog Number: (102830-850)
Supplier:  Janitorial Supplies
Description:   Enjoy a deeper bass experience over a range of music, movies and games with our down-firing subwoofer with a 4" driver. Easily command your sound with...
Supplier:  Janitorial Supplies
Description:   New Piezo technology delivers powerful performance while significantly reducing weight and increasing battery life. Full frequency response provides c...
Catalog Number: (500026-128)
Supplier:  Janitorial Supplies
Description:   The perfect companion for LCD projectors, this portable speaker/amplifier combination is ideal for audiences of up to 1,000 in rooms up to 10,000 sq. ...
Catalog Number: (102830-856)
Supplier:  Janitorial Supplies
Description:   Rich stereo sound with deep bass for any living space there's the Logitech Z200 multimedia 2.0 stereo speakers. With 10 watts of peak power and two dr...
Catalog Number: (500017-274)
Supplier:  Janitorial Supplies
Description:   Handsfree professional cardioid mic offers superior frequency response. Forty-foot cord with twelve-foot extension for wireless or condenser inputs le...
Supplier:  Janitorial Supplies
Description:   The perfect companion for LCD projectors, this portable speaker/amplifier combination is ideal for audiences of up to 1,000 in rooms up to 10,000 sq. ...
Supplier:  Janitorial Supplies
Description:   Ideal for audiences up to 2,500 people and rooms up to 10,000 sq. ft., this portable PA system features a powerful amplifier to fill the room with ric...
Catalog Number: (500041-499)
Supplier:  Janitorial Supplies
Description:   Entertain audiences up to 500 people with this PA system that’s made for sharing a full multimedia experience. Play music and recharge your iPod® or i...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8