Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "mail-sorters-sorting-stations"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   The basic building block of an efficient mailroom system. Sorter features large 11 5/8w x 13 1/4d x 2 5/8h compartments that easily accommodate heavy ...
Product and/or packaging is safer to human and environmental health
Catalog Number: (500025-097)
Supplier:  Janitorial Supplies
Description:   Create a sorter that works for you. Stack up to two Modules on top of Riser or three Modules without Riser. 11" wide plastic mail trays form up to 25 ...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Adjustable to meet your individual needs. Heavy-duty construction ensures exceptional stability. Large shelf provides ample storage space.
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Heavy-gauge steel components. Use individually or in combination. Quick, easy assembly. Baked enamel finish.
Catalog Number: (500000-544)
Supplier:  Janitorial Supplies
Description:   Heavy-gauge steel for strength and stability. Adjusts for standing or seated comfort. Leveling glides for uneven floors. Top and Base sold and shipped...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Catalog Number: (500026-595)
Supplier:  Janitorial Supplies
Description:   It's almost too E-Z! Perfect for all of your mailroom needs. Smooth surface with durable construction. Tough edges prevent daily wear-and-tear. Quick ...
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Keep your mailroom running smoothly. Transparent label holders have self-adhesive backing and fit securely on beveled shelf front. Lets you easily ide...
Product and/or packaging is safer to human and environmental health
Catalog Number: (500024-150)
Supplier:  Janitorial Supplies
Description:   Supports one or more mail sorter modules or use on a table to accomodate scales, postage meters and off surface computer storage. Cable management cut...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next