Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "carts-and-dollies-service-utility"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"10"
Sort by:
 
 
 
 

Catalog Number: (500004-340)
Supplier:  Janitorial Supplies
Description:   3-3/8" tray lip reverses to a flat top.
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Quiet, lightweight and easy to maneuver cart. Two large trays hold items to be transported. 300 lb. capacity. Molded-in handle with handy storage comp...
Supplier:  Janitorial Supplies
Description:   Won't rust, dent, chip or peel. Lightweight for easy maneuverability and quiet operation. No sharp edges to nick walls or furniture. Two-shelf cart wi...
Supplier:  Janitorial Supplies
Description:   Won't rust, dent, chip or peel. Lightweight for easy maneuverability and quiet operation. No sharp edges to nick walls or furniture. Flat shelves prov...
Catalog Number: (500004-525)
Supplier:  Janitorial Supplies
Description:   Large 6 3/4" wheels and metal telescoping tubing make it ideal for transporting boxes, equipment and presentation material. Easily glides over curbs, ...
Supplier:  Janitorial Supplies
Description:   Lightweight and quiet cart easily maneuvers around office. No sharp edges to damage doorways, furniture or walls. Convenient shelves have one flat sid...
Supplier:  Janitorial Supplies
Description:   Sleek, contemporary styling with 20% more top-shelf capacity is ideal for front or back of house applications. Durable 100 lb. capacity per shelf cons...
Supplier:  Janitorial Supplies
Description:   Ideal storage solution for industrial and commercial use. Snap-together design assembles in minutes. Strong welded wire construction provides exceptio...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 10
  1  2  Next