Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "boards-and-accessories-easel-boards"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"12"
Sort by:
 
 
 
 

Catalog Number: (500009-167)
Supplier:  Janitorial Supplies
Description:   Give a great presentation with a versatile dry erase easel. The magnetic surface means you can display memos, graphics and photos. Flip chart retainer...
Catalog Number: (500009-159)
Supplier:  Janitorial Supplies
Description:   When versatility is key, count on this adjustable height easel. Black steel easel adjusts from 40" to 70" for tabletop or floor-standing height. Featu...
Supplier:  Janitorial Supplies
Description:   Just a simple flip doubles your writing space—double-sided easel features two Total Erase® surfaces. Rotate the board for additional writing space on ...
Supplier:  Janitorial Supplies
Description:   Three-in-one design increases productivity—use as a presentation easel to write notes, a display easel to post information or as a rolling room divide...
Supplier:  Janitorial Supplies
Description:   Sleek design is perfectly angled for writing and viewing. Dual-purpose easel provides the option of using either a whiteboard surface or flipchart pad...
Catalog Number: (500009-133)
Supplier:  Janitorial Supplies
Description:   Versatile features offer lots of options for powerful presentations. The double-sided board tilts and locks at any angle to make viewing and writing i...
Catalog Number: (500009-148)
Supplier:  Janitorial Supplies
Description:   Make an impressive presentation with this dual-sided easel. Pad retainers on both sides securely hold 27w x 34h pads (sold separately). Scratch- and d...
Catalog Number: (500009-147)
Supplier:  Janitorial Supplies
Description:   Ultra-sturdy, ultra-smart design for easy setup and transport. Constructed of blow-molded, high-density, dent- and scratch- resistant polyethylene. De...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 12
  1  2  Next