Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "lamps-and-lighting-desk-table"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"13"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Traditional style desk/table lamp with glass shape and brass base. Accepts 60W bulb GEL41028 (sold separately). Overall height 14".
Catalog Number: (500016-420)
Supplier:  Janitorial Supplies
Description:   Shed some light wherever it's needed. Multipurpose clamp attaches lamp to any table top. 30" swing arm. Vented shade allows heat to escape to help mai...
Catalog Number: (500016-385)
Supplier:  Janitorial Supplies
Description:   A high-tech look for the modern office. Adjustable black metal arm with 18" reach. Heavy weighted base for stability.
Catalog Number: (500016-395)
Supplier:  Janitorial Supplies
Description:   A contemporary classic. Wide-profile domed shade distributes light evenly over work area. Compact weighted base requires minimal desktop space. Bulb s...
Catalog Number: (500016-384)
Supplier:  Janitorial Supplies
Description:   Classic design and antique brass finish blend with most office or home décor. Accepts three-way 50/100/150W bulb, GEL41280 (sold separately).
Catalog Number: (500016-410)
Supplier:  Janitorial Supplies
Description:   13W full spectrum desk lamp let’s you see colors accurately and helps reduce the glare that can cause eye strain. Uniquely designed, weighted, swivel ...
Catalog Number: (500016-405)
Supplier:  Janitorial Supplies
Description:   Swing arm, weighted base for stability. “Easy-dial” on/off cap switch. Accepts 75W bulb, GEL41032 (sold separately).
Catalog Number: (500016-421)
Supplier:  Janitorial Supplies
Description:   Helps keep your desktop organized. Contemporary style with adjustable swing arm. 8-3/8 x 5 base provides convenient storage space for desk essentials.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 13
  1  2  Next