Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "labels-media"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Works with both inkjet and laser printers. Create labels for all your diskettes with easy-to-use Avery® online templates and software. 8-1/2 x 11 shee...
Supplier:  Janitorial Supplies
Description:   Perfect for creating a professional-looking label. Print-to-the-edge capability. Two labels per sheet.
Catalog Number: (500015-833)
Supplier:  Janitorial Supplies
Description:   Design and apply labels that make your discs stand out. Print-to-the-edge capability. Special film material designed for DVDs won’t lift, peel or wrin...
Supplier:  Janitorial Supplies
Description:   Easily create and print professional-looking labels for your discs. Covers 100% of the CD, allowing more space for customized logos, graphics and text...
Catalog Number: (500015-787)
Supplier:  Janitorial Supplies
Description:   Easy-to-use applicator for applying CD/DVD labels. Centers label perfectly every time. Tall center spindle prevents label curl.
Catalog Number: (500015-792)
Supplier:  Janitorial Supplies
Description:   Customize CDs/DVDs with labels and inserts that feature print-to-the-edge capability. Jewel case inserts have ultra-fine perforations for easy separat...
Catalog Number: (500015-925)
Supplier:  Janitorial Supplies
Description:   One standard or two slimline inserts per sheet. Print-to-the-edge capability. Ultra-fine perforations for easy separation.
Supplier:  Janitorial Supplies
Description:   Label and jewel case insert on a single sheet makes it easier and faster to design and print. Print on both the front and back of jewel case insert. P...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next