Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "file-folders-hanging"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"38"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Stronger and tougher than other hanging folders. Reinforced rods are 40% stronger to resist bending and paper tearing. Heavy-duty 1/3-cut adjustable o...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Get your files in order with these classics. Lighter color interior helps prevent misfiling papers between folders. 11 pt. stock. Includes index tabs ...
Supplier:  Janitorial Supplies
Description:   Eco-friendly, high-quality construction—low impact on the environment. Polylaminate strips reinforce top edges and bottom fold for extra durability. T...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Vivid colors help you code files for quick and easy identification. Coated rod tips ensure smooth sliding along your drawer's rails. Durable 11 pt. pa...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Catalog Number: (500011-826)
Supplier:  Janitorial Supplies
Description:   Stylish colors to accent home and office or color-code documents. Rod tips are coated for smooth gliding on hanging rails. 11 pt. colored stock. Slott...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Built-in reinforced tabs are attached to the folder—no assembly required. Stronger than ordinary hanging folders and easy to label by writing directly...
Supplier:  Janitorial Supplies
Description:   Hanging folders keep files standing at attention, eliminating shirkers that slump and go AWOL by slipping under their drawer-mates. Coated rod tips en...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Use color to simplify filing: categorize subjects by color and count on the coordinating, lighter interior color to help prevent misfiles. Now include...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 38
  1  2  3  4  5  Next