Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "file-folders-dividers"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Group papers together in sequence for quick reference and easy access. 2" capacity prong fastener holds papers securely to the sturdy file back. Fits ...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Convenient way to include items that should not or cannot be punched when you use a two-prong filing system. Generous pocket will expand to hold elect...
Supplier:  Janitorial Supplies
Description:   Add documents to fastener folders without hole punching them. Perfect for large hospitals, medical or dental offices or any situation where you need t...
Catalog Number: (500042-626)
Supplier:  Janitorial Supplies
Description:   Three sections in one folder help manage multiple documents or subdivide projects. Sections are closed on two sides to prevent papers from falling out...
Catalog Number: (500040-251)
Supplier:  Janitorial Supplies
Description:   Envelope has flap with string tie and button closure to safely store diskettes, CDs, photos and more. Two-hole punched at top for attachment to prong-...
Supplier:  Janitorial Supplies
Description:   The printable Table of Contents coordinates with preprinted tab titles. Now three ways stronger to last longer—stronger heavier weight paper, stronger...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   Subdivide existing conventional end or top tab folders by adding one or more dividers inside with the scored, self-adhesive edge. Hinged edge is slott...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Supplier:  Janitorial Supplies
Description:   For paper intensive end tab or top tab file folders. Allows you to create many filing surfaces for extensive categorization within a single file folde...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8