Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "file-cabinets-lateral-metal"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Counterweight included, where applicable, to meet ANSI/BIFMA stability requirements. Lock secures both sides of drawer. Heavy-duty, three-part, telesc...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Three-part telescoping slide suspension. Lock secures both sides of drawer. Mechanical interlock allows only one drawer to open at a time to inhibit t...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Hinged doors conceal 36 1/4" storage space with two adjustable shelves. Bottom shelves hold two rows of three-ring binders. Keyed alike locks secure d...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Three-part, telescoping, steel ball bearing suspension for smooth drawer operation. Drawers include side-to-side hangrails for letter or legal size fi...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   An excellent choice for a small office or home office setting. Heavy-duty three-part telescoping slide suspension operates on steel ball bearing suspe...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   An affordable lateral file for light-duty use. Locking drawers with ball bearing slide suspension. Side-to-side rails hold letter/legal size files. Me...
Supplier:  Janitorial Supplies
Description:   Makes three rows of front-to-back letter size filing. Order one pack per drawer/shelf. Two rails per pack.
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Pack makes two rows of letter or legal size filing in one drawer/shelf of 36" files, or two drawers of 30" files. Four rails per pack.
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8