Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

desk-accessories-desktop-shelves

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"11"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Because there is no such thing as too much desk space, free up valuable work surface by storing binders, telephone and monitor on this stylish off-sur...
Supplier:  Janitorial Supplies
Description:   Ideal for desktop literature organization. Shelves are adjustable or removable to customize compartments for document, binder and office supply storag...
Catalog Number: (500017-904)
Supplier:  Janitorial Supplies
Description:   Add storage space without obstructing your view. Full-width shelf with adjustable dividers and two 9 x 11 trays for papers that can be mounted on the ...
Catalog Number: (500024-718)
Supplier:  Janitorial Supplies
Description:   Lifts items 7" off desktop. Perfect for card files, CDs, clocks and other supplies.
Supplier:  Janitorial Supplies
Description:   Made of heavy gauge steel construction, these units can be stacked up to three high to customize you storage. Organizes literature, files, stationery....
Catalog Number: (500017-891)
Supplier:  Janitorial Supplies
Description:   This compact multipurpose organizer brings versatility and accessibility for storage in small spaces. Includes two 17w x 11 1/2d shelves. Literature s...
Supplier:  Janitorial Supplies
Description:   This vertical desktop sorter is compact for maximum desktop organization in a minimum of space. Shelves are removable only during assembly so you can ...
Catalog Number: (500024-716)
Supplier:  Janitorial Supplies
Description:   Turn an unused corner into useful storage with this contemporary mesh shelf. Strong enough for binders, catalogs, books. Three-shelf divider keeps boo...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next