Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "credenzas-wood"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"22"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Durable 1" thick top is covered with abrasion- and stain-resistant laminate. Full-height modesty panel and convenient cord management grommets. Use al...
Catalog Number: (500007-052)
Supplier:  Janitorial Supplies
Description:   18th century elegance, 21st century engineering. Mirror-finish, high-pressure laminate top with traditional edge detail. Woodgrain laminate end panels...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   A modular management solution that combines stylish profiles and elegant accents with top-grade fit and finish. Laminate with rounded corners and wate...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Simple curves create an elegant setting for any work environment. Modular and non-handed construction for flexibility and versatility. Veneers are boo...
Supplier:  Janitorial Supplies
Description:   A modular management solution that combines stylish profiles and elegant accents with top-grade fit and finish. High-pressure laminate with rounded co...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Combining veneer elegance with modular format versatility. Fine-finished, premium-grade veneers and solid woods. Provides durability suited to everyda...
Supplier:  Janitorial Supplies
Description:   Sophisticated and elegant. Designed for both food and beverage services and audio/visual applications. AA-grade select hardwood veneer; surfaces are p...
Supplier:  Janitorial Supplies
Description:   Fashionable aesthetics combined with unparalleled quality yet remarkably affordable. Thermally-fused laminate offers exceptional abrasion- and stain-r...
Product and/or packaging is safer to human and environmental health
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 22
  1  2  3  Next