Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "copyholders-desktop"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"19"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Provides a practical, cost effective solution for organizing reference documents into one convenient location. Sturdy metal desk stand with rugged pan...
Catalog Number: (500006-566)
Supplier:  Janitorial Supplies
Description:   Flexible arm adjusts to correct viewing height and angle. Pivoting line guide glides up and down, swings out of the way. Copyholder plate rotates 90° ...
Supplier:  Janitorial Supplies
Description:   Adjustable elastic guideline secures thick documents. The unit holds books, catalogs and landscape sheets, and allows multiple angle and height adjust...
Product and/or packaging contains recycled/renewable content
Catalog Number: (500006-562)
Supplier:  Janitorial Supplies
Description:   Steel mesh copyholder in elegant modern design allows for easy one-handed document attachment. Steel ball securely holds papers and solid metal center...
Product and/or packaging is safer to human and environmental health
Supplier:  Janitorial Supplies
Description:   Unique track design allows line guide and paper holder to be positioned at any location along the top or sides. Easel back adjusts for proper height a...
Catalog Number: (500006-546)
Supplier:  Janitorial Supplies
Description:   Holds both A4 and letter size documents. Adjustable for various viewing angles. Includes paper holder/line guide and pen holder.
Catalog Number: (500006-548)
Supplier:  Janitorial Supplies
Description:   Positioning your documents at a more comfortable viewing angle will not only boost productivity but also help reduce strain on your neck. Features a s...
Catalog Number: (500006-552)
Supplier:  Janitorial Supplies
Description:   Magnetic paper holder keeps copy in place. Folds flat for storage. Features a removable line guide and adjustable reading angle.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 19
  1  2  3  Next