Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "computer-furniture-workstations"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Melamine top reverses during setup for a choice of mahogany or medium oak finish. Black steel base with full-width 6-3/4" deep storage shelf. 12" mode...
Supplier:  Janitorial Supplies
Description:   Scratch- and stain-resistant melamine surface. Steel tube support column has levelers to compensate for uneven flooring. Not freestanding. Combine wit...
Supplier:  Janitorial Supplies
Description:   Versatile split-level design allows table to be configured in several ways. Gray laminate surfaces with black powder coated steel frame base. Leveling...
Supplier:  Janitorial Supplies
Description:   Ultra-modern computer desk holds convenient, retractable keyboard platform. Durable, glass and laminate work surface stands up to dust and dirt elemen...
Catalog Number: (500030-706)
Supplier:  Janitorial Supplies
Description:   Large black laminate CPU bay with locking door for equipment security. Molded plastic keyboard tray with built-in covered pen/pencil holder. Wire chan...
Catalog Number: (500026-491)
Supplier:  Janitorial Supplies
Description:   Folds to 5-1/4" deep for storage and mobility. Drop keyboard shelf with six height and tilt positions. Light gray 3/4" laminate top on medium gray ste...
Supplier:  Janitorial Supplies
Description:   Set up your workstation to suit your ergonomic needs and work style. Unit adjusts to three different heights to maximize viewing comfort and reduce ne...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7