Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "cleaning-supplies-computer-surface-cleaners"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"11"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   JohnsonDiversey Antibacterial Multi-Surface Cleaner 32 oz. Spray Bottle
Catalog Number: (500037-780)
Supplier:  Janitorial Supplies
Description:   Keep your expensive equipment clean and dust-free. Premoistened, antistatic, nonabrasive wipes are safe to use on electronic equipment and delicate su...
Catalog Number: (500039-453)
Supplier:  Janitorial Supplies
Description:   Nontoxic all purpose spray is gentle on the environment yet tough on dirt. Naturally derived formula is safe on most home surfaces including tile, mar...
Catalog Number: (500037-776)
Supplier:  Janitorial Supplies
Description:   Premoistened, antistatic wipes are soft and thick to safely remove dust, dirt and lint from sensitive computer and laptop screens. Quick-drying, no-st...
Supplier:  Janitorial Supplies
Description:   New and improved formula. Premoistened, ammonia-free wipes remove fingerprints, dirt and dust, and antistatic action prevents dust buildup. Convenient...
Supplier:  Janitorial Supplies
Description:   Generously sized, premoistened, alcohol-free wipes quickly and safely remove dust, dirt, grime, fingerprints and smudges from screens. Nonabrasive and...
Catalog Number: (500037-892)
Supplier:  Janitorial Supplies
Description:   Premoistened, nonabrasive, nonstreaking wipes remove dust, fingerprints and smudges from notebook computer screens. Individually wrapped for convenien...
Supplier:  Janitorial Supplies
Description:   One-step solution to cleaning office equipment, desktops and any plastic or metal surface. Presaturated wipes remove dust, dirt, fingerprints, ink and...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next