Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

cleaning-supplies-computer-compressed-air

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Blows away harmful dust and lint. 100% ozone safe; no CFCs, HCFCs, butane or propane. Specially developed for use in environments such as scientific r...
SDS
Supplier:  Janitorial Supplies
Description:   Blows away harmful dust and lint. 100% ozone safe; no CFCs, HCFCs, butane or propane. Trigger controls spray power. Slip-on extender offers greater re...
SDS
Catalog Number: (500042-565)
Supplier:  Janitorial Supplies
Description:   High-performance compressed air duster removes dust and lint from electronics. Flexible extension tube provided for tight spaces and pinpoint cleaning...
Supplier:  Janitorial Supplies
Description:   Safely remove lint and dust from keyboards, computers and more. Includes a 5" wand for those hard-to-reach areas. Guilt-free use because it's 100% ozo...
SDS
Supplier:  Janitorial Supplies
Description:   Prolongs the life of PCs, VCRs, stereos and other valuable electronic equipment. Quickly and safely blows away dust and lint from hard-to-reach places...
SDS
Catalog Number: (500032-806)
Supplier:  Janitorial Supplies
Description:   100% ozone safe. 5" extension wand cleans printer parts, keyboards and diskettes. Disposable. Nonflammable.
Catalog Number: (500032-634)
Supplier:  Janitorial Supplies
Description:   75% stronger than regular dusters. Ideal for tough jobs. 100% ozone safe—contains no CFCs. 5" extension wand for tight spots. Gently removes dust and ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7