Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

cleaning-supplies-scrubbers-and-sponges

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"17"
Sort by:
 
 
 
 

Supplier:  Janitorial Supplies
Description:   Promote cleanliness in your home or business with these sponges. Sponges feature a unique StayFresh antimicrobial technology that resists bacterial od...
Catalog Number: (500025-146)
Supplier:  Janitorial Supplies
Description:   Some cleaning jobs requiring heavy-duty supplies--now you have one. These commercial-sized sponges are ideal for big jobs, when you need a long lastin...
Catalog Number: (500039-369)
Supplier:  Janitorial Supplies
Description:   Scrub sponge head refills keep your dishwand like new. Teardrop shape and scouring pad gets into all the corners, nooks and crannies.
Catalog Number: (500018-511)
Supplier:  Janitorial Supplies
Description:   Open-texture, medium-abrasive green pad for general-purpose cleaning and scouring. High-quality synthetic fiber will not rust or corrode.
Catalog Number: (500018-533)
Supplier:  Janitorial Supplies
Description:   Tackles rough cleaning chores. Cleans pots, pans, grills and ovens. Removes floor scuff marks. Won't rust or splinter.
Supplier:  Janitorial Supplies
Description:   Removes tough, baked-on messes 50% faster than other competitive sponges.
Supplier:  Janitorial Supplies
Description:   Multi-purpose scrub sponge cleans tough messes. Clean cookware, countertops or any other surface with a finish you don’t want to scratch--even wood or...
Supplier:  Janitorial Supplies
Description:   Absorbent cellulose sponge on one side, medium-duty nylon scrub pad on the other. Ideal for cleaning ceramic fixtures and other kitchen and bath surfa...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 17
  1  2  3  Next