Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "1-2-dimethylcyclohexane"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 583-57-3 MDL Number: MFCD00064172 Molecular Formula: C8H16 Molecular Weight: 112.22 Purity/Analysis Method: 98.0% (GC) Form: Clea...