    Sort by:
  • Search Within Results
You Searched for: "4-isopropylcyclohexanol"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (TCI0780-5G)
Supplier:  TCI America
Description:   CAS Number: 4621-04-9 MDL Number: MFCD00043476 Molecular Formula: C9H18O Molecular Weight: 142.24 Purity/Analysis Method: 98.0% (GC) Form: Cl...