GammaGuard CE™ Hoods, Sterile, International ENVIROGUARD™
Supplier: International Enviroguard
CE11063CIS
89127-970CS 678.63 USD
89127-970
GammaGuard CE™ Hoods, Sterile, International ENVIROGUARD™
GammaGuard CE™ sterile cleanroom apparel offers a superior liquid barrier, breathability, low linting, and tear resistance
Full face hoods are white with a tunnelized elastic face. Hoods are sterilized to a 10-6 Sterility Assurance Level (SAL).
Special fold design prevents contamination during gowning. Material contains no natural rubber latex. Recyclable.
Ordering information: Manufactured in an ISO-certified facility, clean processed, and individually packaged under cleanroom conditions.