Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "egta-na4-ethylene-glycol-bis-2-aminoethyl-ether-n-n-n-n-tetraacetic-acid-tetrasodium-salt-in-aqueous-solution"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  VWR
Description:   EGTA, (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid), ultrapure, CAS Number: 67-42-5, Molecular Formula: C₁₄H₂₄N₂O₁₀, MW: 380.4 g/mol, Melting Pt: 240 to 244 deg C, size: 50 g