    Sort by:
  • Search Within Results
Disodium α-glycerophosphate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 1555-56-2 MDL Number: MFCD00035539 Molecular Formula: C3H9O6P Molecular Weight: 216.04 Purity/Analysis Method: 85.0% (T) Form: Cr...