You Searched For: Ethanol


34  results were found

SearchResultCount:"34"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR
Description: ETHANOL 70% BIOTECHNOLOGY GRADE 4 L
Supplier: MilliporeSigma
Description: GR ACS, Meets Reagent Specifications for testing USP/NF monographs
Supplier: MilliporeSigma
Description: Ethanol 96% EMSURE® Reag. Ph Eur.
Supplier: MilliporeSigma
Description: Offering a high degree of UV transmittance, low particle count, low acidity and alkalinity, and low evaporation residue level, LiChrosolv® solvents ar...
Supplier: MilliporeSigma
Description: Emplura* Ethanol absolute, Cas number: 64-17-5, Chemical Formula:C2H5OH, Solvent for lab application, 25l.
Supplier: MilliporeSigma
Description: Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents ran...
Supplier: MilliporeSigma
Description: Ethyl Alcohol, 190 Proof HPLC. Suitable for multiple applications, these reagents meet or exceed ACS specifications.
Supplier: MilliporeSigma
Description: ETHANOL ABSOLUTE FOR ANAL (1L)
Supplier: MilliporeSigma
Description: Grade ACS , Cas Number - 64-17-5
Supplier: Decon Labs
Description: 140 proof pure (undenatured) ethanol (70%) USP. Used for research, healthcare, industrial, and production applications as a cleaner, solvent, and reag...
Supplier: Decon Labs
Description: 190 proof (95%) pure (undenatured) ethanol. Meets USP and multi-compendial specifications. Used for research, healthcare, industrial, and production ...
Supplier: Decon Labs
Description: Pure (undenatured) anhydrous (200 proof/100%) ethyl alcohol meets USP and Multicompendial specifications used as a cleaner, solvent, or reagent.
Supplier: Decon Labs
Description: Decon’s 200 proof ethanol (100% ethanol) is a Molecular Biology Grade, pure ethanol product made to USP specifications and is tested for DNAase / RNAa...
Catalog Number: (71001-754)
Supplier: Decon Labs
Description: ETHANOL 200 PROOF 1 PT

Catalog Number: (76212-358)
Supplier: Decon Labs
Description: This 140 Proof ethanol (70% ethanol) is a pure ethyl alcohol made for research, healthcare, industrial, or production applications as a cleaner, solve...

Supplier: MilliporeSigma
Description: ETHYL ALCOHOL PURE 200PF 4L
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 34
no targeter for Bottom