Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
Flow Controllers
https://us.vwr.com/stibo/smallweb/std.lang.all/52/49/18945249.jpg
A flow controller is a device used to measure and control the flow of liquids and gases. Flow controllers can be either analog or digital, and they are designed and calibrated to control a specific type of liquid or gas at a particular range of flow rates. They are typically operated in the flow range of 10 to 90% for the best accuracy. Both manifold- and single-flow versions are available, and are easy to install and operate.
SearchPresentationType-VERTICAL
SearchResultCount:"3"
  List View Easy View
Sort by:
Supplier:  Wilmad-LabGlass
Description:   These flow controllers control pressure applied to a reservoir or directly to a chromatography column
Supplier:  Corning
Description:   Standard flow controller consists of a bottom 35/20 ball joint and a 0 to 4 mm top valve.
Product available on GSA Advantage®
Supplier:  Restek
Description:   Veriflo® 423XL flow controllers are offered in a Siltek®-treated and stainless steel version.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 3  of 3