Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
Zero Air Generators
https://us.vwr.com/stibo/smallweb/std.lang.all/00/37/18990037.jpg
Zero air generators can provide a source of ultra high-purity (UHP) grade zero air for combustion gas. Systems produce UHP zero air with a purity level below 0.05ppm total hydrocarbon content from a compressed air supply. The air generated can be used as support gas for total hydrocarbon analyzers, as nebulizer and exhaust pump gas for LC/MS instruments, or as oxidants/support gas for GC with FID, FPD, and NPD detectors.The generators are engineered for easy installation, operation, and long-term performance.
SearchPresentationType-VERTICAL
SearchResultCount:"4"
  List View Easy View
Sort by:
Supplier:  VWR International
Description:   Systems produce UHP zero air with a purity level below 0.05ppm total hydrocarbon content from a compressed air supply.
Supplier:  VWR International
Description:   VWR® FID Gas Stations are designed to provide fuel gas and support air to flame ionization detectors, flame photometric detectors, and total hydrocarb...
UL Listed CSA Certified
Supplier:  VWR International
Description:   Reliable, durable generators can provide nitrogen gas and zero air to flame ionization detectors (FID) on gas chromatographs.
Supplier:  Restek
Description:   For use with Parker Balston® zero air generators.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 4  of 4