Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
You Searched For:
Soxhlet and Soxhlet-Type Extractors
20  results were found
https://us.vwr.com/stibo/smallweb/std.lang.all/98/18/18989818.jpg
Used when compounds have limited solubility, glass Soxhlet extractors successfully separate lipids from solid material. With the temperature resistance needed to heat solvents, the specialized apparatuses allow vapors to travel up the arm where a thimble blocks solid material from escaping. Designed to have cooling vapor enter the solid contained chamber, percolators circulate the now warm solvent liquid to dissolve desired compounds. Incorporated vapor tubes protect the bulbed siphon tube within Soxhlet extractor systems.
SearchPresentationType-VERTICAL
SearchResultCount:"20"
  List View Easy View
Sort by:
Supplier:  Glassco
Description:   Extractor body only.
Catalog Number: (KT617600-0025)
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Primarily used to determine fat content in dairy products but may also be used for other food products.
Supplier:  Wilmad-LabGlass
Description:   These borosilicate glass extraction apparatus are used to measure the fat content in dairy and other food products.
Supplier:  Corning
Description:   For use with PYREX® extraction apparatus (see 27613-022 series) with [ST] ground joints at top and bottom
Product available on GSA Advantage®
Supplier:  Corning
Description:   Extraction apparatus consists of an extraction tube, a special bulb-type condenser, and a flask
Product available on GSA Advantage®
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Extraction apparatus consists of an Allihn condenser, flask and Soxhlet extraction tube
Supplier:  Wilmad-LabGlass
Description:   These borosilicate glass Soxhlet extraction apparatus are used for the continuous extraction of solids with a suitable solvent
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Used for the continuous extraction of solids with a suitable solvent
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 20
  1  2  3  Next