Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • MWBE (116) SBE (103)
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

pliers

https://us.vwr.com/stibo/smallweb/std.lang.all/96/58/18989658.jpg

With an increase of efficiency, the handheld pliers clamp and hold objects firmly or can even manipulate difficult materials. Constructed of strong steels and additives to improve strength and corrosion resistance, the serrated jaws easily press together in a magnified force by squeezing the cushioned handle levers together. Different configurations allow for positioning and handling of minute, hazardous, or awkward-to-reach objects in any field. High-grade pliers can be autoclaved for clean surgical purposes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"167"
Sort by:
 
 
 
 

Supplier:  MicroSolv Technology Corp
Description:   The Beta Tool™ is a pair of precision, high grade pliers and can be used as needle nose pliers or as a tubing straightener.
Catalog Number: (10195-558)
Supplier:  Sklar
Description:   Plier, 5 1/2inch Square End Flat Nose, Straight, Cross Serrated, Blunt, Stainless Steel, Reusable, Non-Sterile, Latex-Free, Premium OR Grade
Supplier:  Sklar
Description:   College Cotton Pliers are construced of stainless steel. Industry standard. This thumb instrument most commonly used to insert and remove cotton or ot...
Supplier:  Sklar
Description:   Merit™ Meriam Cotton Pliers are stainless steel and industry standard.
Supplier:  Sklar
Description:   Goslee Flat Nose Pliers are stainless steel and industry standard.
Catalog Number: (10195-534)
Supplier:  Sklar
Description:   Slip Joint Pliers are constructed of stainless steel. Industry standard.
Supplier:  Sklar
Description:   Sklar® London College Dental Cotton Pliers #317A are a thumb instrument most commonly used to insert and remove cotton or other dressing materials dur...
Catalog Number: (10193-334)
Supplier:  Sklar
Description:   Needle Nose Pliers are a non-ratcheted, plier handle instrument used for grasping and manipulating k-wire. They are used to guide and twist the k-wire...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 167
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next