Rubbermaid® Commercial Round Storage Containers
Supplier: Rubbermaid
RCP572024CLE RCP5724WHI RCP572424CLE RCP572924CLE RCP572324CLE
89073-290EA 9.81 USD
89073-290 89073-720 89073-302 89073-296 89073-294
Rubbermaid® Commercial Round Storage Containers
Economical, ultra-durable containers fulfill a wide variety of foodservice and general storage requirements.
  • Stackable handled containers made of durable polyethylene
  • Temperature range: –34 to 100 °C (–20 to 212 °F)
  • Dishwasher safe
Easy-grip handles provide better control when pouring contents. Reinforced rim enhances rigidity, preserving the opening's symmetry. Stackable, nesting design enables space-saving storage.
Certifications: SNSF 3-A And NSF 2 Certified.