VWR® Talon® Heavy-Duty Pinchcock
915040
21573-650CS 1195.62 USD
21573-650
VWR® Talon® Heavy-Duty Pinchcock
Handle tough jobs easily with this pinchcock and heavy-duty spring.
  • Simple, one-handed operation
  • Ideal for start/stop precision
  • Resistant to corrosion and rust
Choose it when using stiff tubing common in gas and water connections or when you need more pressure. Offers simple, one-handed operation and quick, convenient, complete closure when placed on tubing and released. Won't cut tubing. Corrosion- and rust-resistant. For heavy-wall tubing up to 11 mm (7/16") O.D.