J.T.Baker®, Spill Clean-Up Centers and Kits
Supplier: Avantor
4437-02 4442-02 BAKR4442-02 BAKR4437-02
JT4437-2CS 2135.56 USD
JT4437-2 CAJT4442-2 CAJT4437-2 JT4442-2
J.T.Baker®, Spill Clean-Up Centers and Kits
Spill Control Materials
Be prepared for hazardous laboratory spills. With NEUTRASORB® and SOLUSORB®, lab personnel can quickly neutralize, clean up, and safely dispose of acid and flammable solvent spills.
  • To neutralize, clean up, and safely dispose of acid and flammable solvent spills
  • NEUTRASORB provides rapid, controlled neutralization of concentrated acids, and visible color change as indication of complete neutralization
  • SOLUSORB reacts immediately with the solvent, reducing solvent vapors and raising the flash point of the mixture
  • Supplied with the appropriate neutralizer, safety glasses, scoops, gloves, disposal bags and ties, and labels replacement kits, available separately
For acid spill clean-up, NEUTRASORB® provides rapid, controlled neutralization of concentrated acids, and visible color change as indication of complete neutralization
For solvent spill clean-up, SOLUSORB® reacts immediately with the solvent, reducing solvent vapors and raising the flash point of the mixture. SOLUSORB® provides for the safe, effective handling of a solvent spill through reduction of flammable or toxic vapors, fast handling, complete cleanup, and convenient disposal.
After complete neutralization, waste materials contain neutralized alkali metal and alkaline earth salts of the treated acid.
Caution: NEUTRASORB® should not be used for spills of hydrofluoric acid, fuming nitric acid, fuming sulfuric acid, or peroxy (per-) organic or other highly reactive acids.