VWR® 384-Well Plates
P9651-VWR P9650-VWR P9757-VWR
89106-790CS 385.54 USD
89106-790 82004-672 89106-792
VWR® 384-Well Plates
Polypropylene storage plates have square wells with a conical base that aids in sample concentration, reconstitution, and centrifugation.
  • For high-throughput screening applications
  • Polypropylene fabrication
The corners of all the wells have a small radius to prevent sample precipitation and improve concentration
The four-compartment freezing rack (89000-622) holds 52 standard plates or 16 deep well plates. The six-compartment freezing rack (89000-624) holds 102 standard or 30 deep well plates.
Designed to meet proposed SBS specifications. Plates are DNase- and RNase-free.
Ordering information: Sterile plates are supplied in inner bags of five plates.