Buchi® R-210 Rotavapor® Evaporators
23022A110 23021C121 23022A000 23023V120 23021V122 23023C120 23021V121 23022A121 23022C120 23022A012 23023C100 23023A100 23021V101 23022C101 23022A022 23023V112 23022A010 23023C110 23023A012 23022V110 23021C110 23022A001 23023C101 23022C110 23022A112 23022V111 23022V120 23021C101 23023A112 23022C112 23022C100 23022A011 23022V122 23022A120 23023V101 23022C111 23023A000 23023A121 23023A110 23023V111 23023A022 23023V100 23023A021 23021V100 23021C112 23023A011 23023A120 23023A010 23022C121 23022A101 23023C121 23021C122 23023C111 23023V122 23023A020 23022V100 23022A021 23023C112 23023A101 23023A111 23023C122 23021V111 23023A122 23022A122 23022A020 23022A100 23022V121 23023V110 23021C120 23021C100 23022A111 23023A001 23022C122 23021V120 23021V110 23022V101 23023V121 23021V112 23022V112 23021C111
89036-248EA 13468.93 USD
89036-248 89036-204 89036-126 89036-246 89036-202 89036-168 89036-124 89036-208 89036-206 89036-128 89036-240 89036-162 89036-160 89036-244 89036-200 89036-166 89036-122 89036-242 89036-164 89036-280 89036-138 89036-258 89036-214 89036-136 89036-212 89036-256 89036-218 89036-216 89036-174 89036-130 89036-250 89036-172 89036-134 89036-178 89036-254 89036-210 89036-132 89036-176 89036-252 89036-170 89036-226 89036-148 89036-268 89036-224 89036-146 89036-228 89036-262 89036-140 89036-184 89036-260 89036-182 89036-266 89036-222 89036-188 89036-144 89036-264 89036-220 89036-186 89036-142 89036-180 89036-236 89036-158 89036-234 89036-278 89036-238 89036-152 89036-196 89036-272 89036-150 89036-194 89036-270 89036-156 89036-276 89036-232 89036-154 89036-198 89036-274 89036-230 89036-192 89036-190
Buchi® R-210 Rotavapor® Evaporators
This series of rotary evaporators provides a complete solution for laboratory solvent evaporation processes by integrating a vacuum controller, vacuum pump, and recirculating chiller into one, compact, safe, reliable, easy-to-use, and maintenance-free system.
Evaporator units are available in basic, advanced, or professional styles, based upon the incorporation of vacuum controllers
Evaporators with diagonal condensers can be used in areas of limited space or height. They are designed for standard distillations and feature a stopcock feed to the evaporating flask. Evaporators with vertical condensers can be used in areas of unobstructed space and height. Designed for standard distillations, they feature a stopcock feed to the evaporating flask, and they provide a connection for vapor temperature sensors. Vertical condensers permit auto distillation with the use of a stage probe and V-855 vacuum controller. Evaporators with dry ice condensers permit the distillation of solvents with low boiling points. They feature a stopcock feed to the evaporating flask and do not require cooling water. Maximum condensation will result due to low temperatures.
Standard features on any Rotavapor® system include continuous vapor ducting to prevent cross-contamination, the option of P+G plastic coating on all glass components to protect against mechanical damage, and a 4L water or oil heating bath with a temperature range of 20 to 180°C (68 to 356°F). A large display continuously shows actual and setpoint temperature. A cordless power supply eliminates the need to unplug connections when filling or emptying the bath, and the small volume, stainless steel bath ensures quick heating capabilities. A quick-action jack, easily operated by the touch of a button automatically raises the flask from the heating bath to prevent thermal loading in the event of a power failure.
Basic evaporators operate without a vacuum controller. Advanced units are supplied with V-850 vacuum controllers, which offer pressure regulation to a setpoint, a library containing 43 commonly-used solvents (with the ability to add your own), a timer function, USB interface for data transfer, and control of quick-action jack and Rotavapor® rotation. The more versatile professional units feature the V-855 vacuum controller, which offers EasyVac functions, automatic distillation, pressure gradient programming, and repeatable distillation functions in addition to the features of the V-850. The V-855 vacuum controller is designed to work optimally in conjunction with V-700 and V-710 vacuum pumps. Process parameters are always visible at a glance due to the direct fastening of all vacuum controllers to the evaporator. The intuitive operating concept makes all unit styles suitable for a wide range of applications.
Certifications: CSA certified. CE marked.