Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Micro-Stir® and Biostir® Magnetic Stirrers, Wheaton

Supplier: DWK Life Sciences
BioStir®
W900703-A W900701-A W900702-A W900700-A
Designed for a variety of cell culture applications and for use with suspension culture flasks

Both the Micro-Stir and the Biostir stirrers have three computer-controlled modes of operation to optimize growth of cells. The constant speed mode is useful for suspension cultures and features a soft start to a constant stirring speed. The interval start mode can be used for attaching cells to microcarrier beads. This mode turns the stirrer on and off using the soft start and stop for a number of cycles, after which the stirrer runs at a constant speed which may differ from the speed used to attach cells. The two-speed stirring mode, also useful for microcarrier applications, slowly starts to a set speed, holds the speed for a user-defined period, and then softly changes to a final constant speed.

Micro-Stir stirrers are ideal for cell culture applications requiring exceptionally slow speeds, gentle mixing, and low heat transfer. The gradual stop rotation facilitates bead and cell attachment by allowing cells to settle and attach and to draw off and replenish any spent media.

Biostir stirrers feature a powerful alnico magnet for increased efficiency and a smooth-running DC motor that provides uniform speed while reducing heat transfer to the medium.

A DC motor and an easy-to-read display provide precise speed control for reproducible results. Units can be used in environments with CO2 and non-condensing humidity.

The remote control allows all stirrer functions to be controlled and monitored without opening an incubator and disturbing the incubator contents. The remote control comes with a 3.7m (12') cord.

Ordering information: Culture flasks sold separately. For Wheaton Celstir® suspension culture flasks, see 62401-900 series.



 

ORDER

Expand All / Collapse All

Micro-Stir Slow Speed Stirrers
No. of Positions Speed Range Supplier No. VWR Catalog Number Unit Price Quantity
1 5–175 rpm W900700-A 12000-906 Case of 1 Retrieving
Small Business Enterprise
4 5–175 rpm W900701-A 12000-908 Case of 1 Retrieving
Small Business Enterprise

Biostir Heavy-Duty Stirrers
No. of Positions Speed Range Supplier No. VWR Catalog Number Unit Price Quantity
1 150–1200 rpm W900702-A 12000-910 Case of 1 Retrieving
Small Business Enterprise
4 150–1200 rpm W900703-A 12000-912 Case of 1 Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Dimensions 20.3W x 24.8D x 8.9H cm (8 x 93/4 x 31/2") 1-Position Stirrer,
39.4W x 44.5D x 9.5H cm (151/2 x 171/2 x 33/4") 4-Position Stirrer
Electrical 115V, 50/60Hz, 15W
Max. Temperature 50°C (122°F)

ACCESSORIES

Expand All / Collapse All

Accessories
No. of Positions Supplier No. VWR Catalog Number Unit Price Quantity
Remote Control W900704 12000-914 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty