Brownlee™ Validated™ HPLC Columns (80 and 100 Å Pore Size), PerkinElmer
Supplier: PerkinElmer
Brownlee™ Validated™
N9303582 N9303582
97019-500EA 1115.45 USD
CA97019-500 97019-500
Brownlee™ Validated™ HPLC Columns (80 and 100 Å Pore Size), PerkinElmer
Chromatography Columns
These columns are packed with high-purity Type B silica with inherent low silanophilic activity.
  • Fine pore design
  • Silica-based sorbent
  • Inherent low silanophilic activity
They exhibit excellent peak shape for a wide range of compounds, while also having a wide number of phases.