Lindberg/Blue M Box Furnaces, 1500 °C, Thermo Scientific
Supplier: Thermo Scientific
BF51333C
30712-055EA 17655.69 USD
30712-055
Lindberg/Blue M Box Furnaces, 1500 °C, Thermo Scientific
Provides superb temperature uniformity through the use of silicon carbide heating elements positioned horizontally in the chamber
Internal controllers display both the actual chamber temperature and setpoint simultaneously on an LED display. The 300-segment programmable controllers feature 30 programs with ramps and dwells. The 16-segment programmable controllers feature one program for ramps and dwells.
Chamber is constructed of Moldatherm® ceramic insulation for rapid heat-up and cool-down, energy efficiency, and cooler exterior surface temperatures. Horizontal door model (30712-055) utilizes brick sidewall composition instead of Moldatherm insulation. Insulating vestibule and tight-fitting insulating door plug provide better temperature uniformity by minimizing heat loss around the furnace chamber opening. An internal vent in the vestibule aids in dissipating the initial heat surge that occurs when the door is opened. Insulating door plug serves as loading platform when in open position. Electrical: 208/240V, 50/60Hz.
Ordering information: Furnaces feature a one-year parts and labor warranty. Furnaces include Type-R thermocouple and hearth plate. Power cords and interconnecting wiring not included. Furnace 30604-215 features over-temperature control that shuts off power if high temperature limit is reached. Furnace 30712-055 includes 3m (10') compensated thermocouple lead wire and polarized plug (requires external controller, sold separately). Optional RS-485 data connection port is available on all models; contact your VWR sales representative.