Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FIBERLite Centrifuge Rotors, Thermo Scientific

Supplier: Thermo Scientific
096-029027 096-145017 096-087051 096-084075 096-124022 096-085065 096-309012 78500 096-064025 096-309015 096-062114 096-062038 096-484020 096-489011
Carbon fiber composite rotors weigh half that of equivalent aluminum rotors

  • Lightweight
  • Fast acceleration and deceleration
  • 65% carbon fiber

Rotors are 65% carbon fiber (embedded in inert epoxy resin) for increased strength, low weight, and high resistance to corrosion, heat, fatigue, and other environmental influences. Insulative properties eliminate necessity of precooling step.

Rotors are supplied with factory-installed centrifuge-specific hub for maximum compatibility with most centrifuges. Rotors accept standard bottles and tubes as used in equivalent metallic rotors. They can be washed and soaked in a wide variety of laboratory detergents, and are autoclavable, cold room compatible, and highly resistant to radiolabel contamination.

Light weight allows for easier handling and reduced risk of lower back injury due to lifting. Rotors also reduce wear and tear on centrifuge drive system and decrease maintenance costs. Faster acceleration and deceleration make total run times 40% shorter for larger rotors.

Ordering information: Rotors are supplied with manufacturer's limited lifetime warrranty.



 

ORDER

Expand All / Collapse All
Description For Use With Maximum RCF Maximum Speed Supplier No. VWR Catalog Number Unit Price Quantity
14 x 50 mL Round Bottom Tube Rotor for Sorvall Super Speed Centrifuges RC4 (USA), Multifuge 4 (Europe) 17,136 g 10,000 rpm 096-145017 97000-216 Each Retrieving
30 x 1.5/2 mL Microtube Rotor for Tabletop Carbon Fiber Centrifuges Eppendorf 5810/5804 (will replace F-45-30-11/FA-45-30-11) 22,172 g 14,000 rpm 096-309012 97000-318 Each Retrieving
30 x 1.5/2 mL Microtube Rotor for Tabletop Carbon Fiber Centrifuges Eppendorf 5417c (will replace F-45-30-11/FA-45-30-11) 22,172 g 14,000 rpm 096-309015 97000-316 Each Retrieving
48 x 1.5/2 mL Microtube Rotor for Sorvall* Super Speed Centrifuges RC2, RC5, RC6, RC26, Evolution RC 46,428 g 21,000 rpm 096-484020 97000-212 Each Retrieving
6 x 100 mL Oak Ridge Bottle Rotor for Sorvall Super Speed Centrifuges RC2, RC5, RC6, RC26, Evolution RC 43,904 g 20,000 rpm 096-064025 97000-226 Each Retrieving
6 x 250 mL Oak Ridge Bottle Rotor for Sorvall Super Speed Centrifuges RC2, RC5, RC6, RC26, Evolution RC (will replace GSA/SLA-1500/SLC-1500) 30,074 g 14,000 rpm 78500 97000-234 Each Retrieving
6 x 500 mL Oak Ridge Bottle Rotor for Beckman High Speed Centrifuges Avanti Series (J/J-E), (will replace JLA-10.500) 17,696 g 10,000 rpm 096-062114 97000-260 Each Retrieving
8 x 39 mL Oak Ridge Tube Rotor for Ultra Speed Carbon Fiber Centrifuges Beckman L Series; Sorvall OTD, Discovery, Ultra Pro 80 Series (will replace Beckman Type 55.2 Ti, Type 50.2 Ti/Sorvall T-1250) 257,600 g 50,000 rpm 096-087051 97000-372 Each Retrieving
8 x 50 mL Conical Tube Rotor for Tabletop Carbon Fiber Centrifuges C3i (will replace AC 50.10A) 16,773 g 12,000 rpm 096-085065 97000-336 Each Retrieving
Rotor for Swinging Bucket Centrifuge GS-6, Allegra, and GP Series (will replace G.H. 3.8) 1938 g 3200 rpm 096-029027 97000-308 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
50 mL Conical to 15 mL Millipore™ Filtration Device Adaptor 010-1340 89218-326 Pack of 2 Retrieving
50 mL Conical to 15 mL Oak Ridge Rotor Adaptor 010-0376 97000-352 Pack of 2 Retrieving
50 mL Conical to 250 mL Round Bottom Rotor Adaptor 75100136 89218-316 Pack of 2 Retrieving
50 mL Conical to 50 mL Round Bottom Rotor Adaptor 010-0377 97000-350 Pack of 2 Retrieving
50 mL Round Bottom to 15 mL Oak Ridge Rotor Adaptor 010-0382 97000-354 Pack of 2 Retrieving
5 x 50 mL Conical to 250 mL Round Bottom Rotor Adaptor 75101073 89218-320 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: Thermo Scientific provides several service options such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Centrifuge Technical Service" in the VWR.com search field.

In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty