VWR® High Throughput Homogenizers
930140
12621-164EA
7695.26
USD
12621-164
VWR® High Throughput Homogenizers
These homogenizers are designed for high-throughput sample processing in microplate, vial set format, or any matrix that can fit in to the 10.2x12.7x6.4 cm (4x5x21/2") easy load tray.
- Ability to process small quantities
- Eliminates cross-contamination
- Rapid sample processing
- Now with Best-in-Class, Exclusive 5-Year Warranty
They produce effective homogenization of hard tissue samples, including animal tissue, seeds, tubers, leaf punches, soil and sediment samples, insects, and microbial cultures
They are also suitable for isolation of protein, DNA, RNA, viruses, and other biological components released during homogenization, as well as for dry sample pulverization for chemical analysis and solubility studies. The shaker produces a reciprocal motion optimized for grinding difficult samples, and the high speed linear motion processes samples quickly. In most cases, samples are processed in two minutes or less. Homogenizers feature an easy-to-load tray that securely locks the microplate or vial set in place, analog controls, and an electronic timer. Homogenized samples can be handled manually or in an automated system.
Grinding vial sets combine the benefits of the microplate format with a small vial size to provide a tool for higher throughput sample grinding. Each set includes chemical-resistant polyethylene or rugged polycarbonate vials, each with a threaded cap and one stainless steel grinding ball (set 12621-172 includes two balls per vial), and a plastic case with a foam well rack. With the homogenizer, the 24-well vial set can pulverize corn kernels into dry powder in 2 minutes; while the 5-well vial set can grind pooled or larger samples, such as 15 lyophilized corn kernels, for field trials.
Grinding balls are used to bead beat plant tissues, seeds, and animal tissues. Balls are made of 440C tarnish-resistant stainless steel and can be retrieved with a magnet. Balls are treated prior to packaging to remove residual oils and contaminants. Balls with 9.5mm (3/8") diameter are used with 4 mL vials to homogenize seeds and larger samples of animal tissue. Balls with 11.1mm (7/16") diameter are used with 15 mL vials to grind larger substances, such as pooled seeds. Balls with 4mm (5/32") diameter are used with deep well plates to homogenize leaf punches and 50 mg or less of animal tissue.
Magnet pipet tips remove stainless steel grinding balls from vials or deep well plates. They are made to fit most pipets with ≤200µL capacity.
The ball dropper/bead dispenser can be used to add 4mm (5/32") stainless steel balls to deep well plates, and to dispense small grinding beads into empty micro-well plates. Consists of ball dropper, reservoir tray, grinding bead swipe card, and plastic retaining sheet.
The ball dropper/bead dispenser can be used to add 4mm (5/32") stainless steel balls to deep well plates, and to dispense small grinding beads into empty micro-well plates. Consists of ball dropper, reservoir tray, grinding bead swipe card, and plastic retaining sheet.
Certifications: RoHS certified. CSA listed. 230V units are CE marked.
Ordering information: Homogenizers are supplied with a 2.3m (71/2') detachable, three-wire cord and plug. All other accessories must be purchased separately. 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart