HEA and PPA HyperCel® Mixed-Mode Chromatography Resins, Pall® Life Sciences
Supplier: Cytiva
20250-026 20260-025 20250-033 20260-030
89047-148EA 189.89 USD
89047-148 89047-158 89047-156 89047-150
HEA and PPA HyperCel® Mixed-Mode Chromatography Resins, Pall® Life Sciences
Resins are based on a combination of electrostatic and hydrophobic properties of the protein and ligands.
  • Based on a combination of electrostatic and hydrophobic properties
  • Industry-scalable
HEA HyperCel™ resin has an aliphatic (n-hexylamine) ligand which has lower hydrophobicity than PPA. PPA HyperCel™ resin has an aromatic (phenylpropylamine) ligand that is highly hydrophobic.
The unique industry-scalable resins are designed for protein capture and impurity removal in a biopharmaceutical environment and provide different selectivities not accessible with traditional ion exchange or HIC methods.